missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Methoxyethyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5527806

 View more versions of this product

Catalog No. AAH5527806



Specifications

Specifications

1-(2-Methoxyethyl)piperazine
87°C (189°F)
193°C to 194°C
13484-40-7
MFCD00191214
105098
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-(2-methoxyethyl)piperazine
2734638
97%
0.97
1.473
5g
C7H16N2O
UN3267
1-2-methoxy-ethyl-piperazine
COCCN1CCNCC1
144.218
144.22

Chemical Identifiers

13484-40-7
144.218
BMEMBBFDTYHTLH-UHFFFAOYSA-N
2734638
COCCN1CCNCC1
C7H16N2O
MFCD00191214
1-2-methoxy-ethyl-piperazine
1-(2-methoxyethyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents