Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2'-Methoxybiphenyl-2-yl)-N-methylmethylamine, 98%

$91.91 - $329.41

Chemical Identifiers

Chemical Name or Material 1-(2'-Methoxybiphenyl-2-yl)-N-methylmethylamine
CAS 902836-95-7
Molecular Formula C15H17NO
Formula Weight 227.31
MDL Number MFCD08064250
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5076603

  SDS
alfa aesar™
H5076603
1g Each for $329.41

AAH50766MD

  SDS
alfa aesar™
H50766MD
250mg Each for $91.91
Description & Specifications

Specifications

Formula Weight 227.31
Chemical Name or Material 1-(2'-Methoxybiphenyl-2-yl)-N-methylmethylamine
Assay 98%
Molecular Formula C15H17NO
MDL Number MFCD08064250
CAS 902836-95-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.