missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%

Manufacturer:  Alfa Aesar™ H5479203

 View more versions of this product

Catalog No. AAH5479203



Description

Description

Applications
1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

Solubility
Soluble in Chloroform.

Notes
Store at 4°C. Keep away from oxidizing agents.
Specifications

Specifications

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole
1g
C11H12N4O4S
4328921
Soluble in Chloroform.
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=NC(=N2)[N+](=O)[O-])C
296.301
296.3
134°C to 139°C
74257-00-4
MFCD00009754
1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole
SFYDWLYPIXHPML-UHFFFAOYSA-N
3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
716901
≥99%

Chemical Identifiers

74257-00-4
296.301
SFYDWLYPIXHPML-UHFFFAOYSA-N
716901
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=NC(=N2)[N+](=O)[O-])C
C11H12N4O4S
MFCD00009754
1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole
3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : No

Recommended Storage : Ambient temperatures

Documents