missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Mesitylene)-1,3-butanedione, TCI America™

$325.42 - $1,066.41

Chemical Identifiers

CAS 6450-57-3
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00477603
InChI Key MBZZDUHWYNESIY-UHFFFAOYSA-N
Synonym 1-(2,4,6-Trimethylbenzene)-1,3-butanedione, (2,4,6-Trimethylbenzoyl)acetone
PubChem CID 367102
IUPAC Name 1-(2,4,6-trimethylphenyl)butane-1,3-dione
SMILES CC1=CC(=C(C(=C1)C)C(=O)CC(=O)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12725G View Documents TCI America
M12725G
5g Each for $325.42
 
M127225G View Documents TCI America
M127225G
25g Each for $1,066.41
 
Specifications

Chemical Identifiers

6450-57-3
204.269
MBZZDUHWYNESIY-UHFFFAOYSA-N
367102
CC1=CC(=C(C(=C1)C)C(=O)CC(=O)C)C
C13H16O2
MFCD00477603
1-(2,4,6-Trimethylbenzene)-1,3-butanedione, (2,4,6-Trimethylbenzoyl)acetone
1-(2,4,6-trimethylphenyl)butane-1,3-dione

Specifications

172°C
45°C
6450-57-3
MFCD00477603
MBZZDUHWYNESIY-UHFFFAOYSA-N
1-(2,4,6-trimethylphenyl)butane-1,3-dione
367102
Crystal-Powder at 20°C
1-(2-Mesitylene)-1,3-butanedione
White-Yellow
5g
C13H16O2
1-(2,4,6-Trimethylbenzene)-1,3-butanedione, (2,4,6-Trimethylbenzoyl)acetone
CC1=CC(=C(C(=C1)C)C(=O)CC(=O)C)C
204.269
204.27
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No