Please login to your online account to display your discounted pricing

1-(2-Isocyanatophenyl)piperidine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(2-Isocyanatophenyl)piperidine
CAS 892501-88-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H14N2O
Formula Weight 202.26
MDL Number MFCD08690238
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC19906DA

  SDS
fisher scientific company
CC19906DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.26
Quantity 1g
Assay 97%
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-(2-Isocyanatophenyl)piperidine
CAS 892501-88-1
Molecular Formula C12H14N2O
MDL Number MFCD08690238
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Assay Percent Range 97%