missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Hydroxyethyl)pyrrole, TCI America™

$177.22 - $658.48

Chemical Identifiers

CAS 6719-02-4
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00191451
InChI Key ZIOLCZCJJJNOEJ-UHFFFAOYSA-N
PubChem CID 10975438
IUPAC Name 2-pyrrol-1-ylethanol
SMILES C1=CN(C=C1)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08261G View Documents TCI America
H08261G
1g Each for $177.22
 
H08265G View Documents TCI America
H08265G
5g Each for $658.48
 
Specifications

Chemical Identifiers

6719-02-4
111.14
ZIOLCZCJJJNOEJ-UHFFFAOYSA-N
2-pyrrol-1-ylethanol
C6H9NO
MFCD00191451
10975438
C1=CN(C=C1)CCO

Specifications

Undesignated
6719-02-4
MFCD00191451
C1=CN(C=C1)CCO
111.14
111.14
≥99.0% (GC)
1g
C6H9NO
ZIOLCZCJJJNOEJ-UHFFFAOYSA-N
2-pyrrol-1-ylethanol
10975438
Clear Liquid at 20°C
1-(2-Hydroxyethyl)pyrrole
Safety and Handling

Safety and Handling

TSCA : No