missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Hydroxyethyl)-3-methylimidazolium Tetrafluoroborate, TCI America™

$96.25 - $331.53

Chemical Identifiers

CAS 374564-83-7
Molecular Formula C6H11BF4N2O
Molecular Weight (g/mol) 213.971
MDL Number MFCD12761438
InChI Key KLTUZFZUYLXTFF-UHFFFAOYSA-N
PubChem CID 11344909
IUPAC Name 2-(3-methylimidazol-3-ium-1-yl)ethanol;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[N+]1=CN(C=C1)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H149225G View Documents TCI America
H149225G
25g Each for $331.53
 
H14925G View Documents TCI America
H14925G
5g Each for $96.25
 
Specifications

Chemical Identifiers

374564-83-7
213.971
KLTUZFZUYLXTFF-UHFFFAOYSA-N
2-(3-methylimidazol-3-ium-1-yl)ethanol;tetrafluoroborate
C6H11BF4N2O
MFCD12761438
11344909
[B-](F)(F)(F)F.C[N+]1=CN(C=C1)CCO

Specifications

Yellow
374564-83-7
MFCD12761438
KLTUZFZUYLXTFF-UHFFFAOYSA-N
2-(3-methylimidazol-3-ium-1-yl)ethanol;tetrafluoroborate
11344909
Clear Liquid at 20°C
1-(2-Hydroxyethyl)-3-methylimidazolium Tetrafluoroborate
25g
C6H11BF4N2O
1760
[B-](F)(F)(F)F.C[N+]1=CN(C=C1)CCO
213.971
213.97
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

TSCA : No