missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Hydroxyethyl)-3,3-dimethylindolino-6′-nitrobenzopyrylospiran 93.0+%, TCI America™

$213.97 - $668.22

Chemical Identifiers

CAS 16111-07-2
Molecular Formula C20H20N2O4
Molecular Weight (g/mol) 352.39
MDL Number MFCD00369031
InChI Key FGJPKPQMQFZPJY-UHFFFAOYSA-N
Synonym 1′C-(2-Hydroxyethyl)-3′C,3′C-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2′C-indoline]
PubChem CID 3104858
IUPAC Name 2-(3',3'-dimethyl-6-nitrospiro[chromene-2,2'-indole]-1'-yl)ethanol
SMILES CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC(=C4)[N+](=O)[O-])CCO)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10421G View Documents Tci America
H10421G
1g Each for $213.97
 
H10425G View Documents Tci America
H10425G
5g Each for $668.22
 
Specifications

Chemical Identifiers

16111-07-2
352.39
FGJPKPQMQFZPJY-UHFFFAOYSA-N
3104858
CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC(=C4)[N+](=O)[O-])CCO)C
C20H20N2O4
MFCD00369031
1′C-(2-Hydroxyethyl)-3′C,3′C-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2′C-indoline]
2-(3',3'-dimethyl-6-nitrospiro[chromene-2,2'-indole]-1'-yl)ethanol

Specifications

Purple-Red
1g
C20H20N2O4
1′C-(2-Hydroxyethyl)-3′C,3′C-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2′C-indoline]
CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC(=C4)[N+](=O)[O-])CCO)C
352.39
352.39
≥93.0% (HPLC,T)
165°C
16111-07-2
MFCD00369031
FGJPKPQMQFZPJY-UHFFFAOYSA-N
2-(3',3'-dimethyl-6-nitrospiro[chromene-2,2'-indole]-1'-yl)ethanol
3104858
Crystal-Powder at 20°C
1-(2-Hydroxyethyl)-3,3-dimethylindolino-6′-nitrobenzopyrylospiran
Safety and Handling

Safety and Handling

TSCA : No