missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 97%, Maybridge

Chemical Identifiers

CAS 6921-64-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
InChI Key LYKDOWJROLHYOT-UHFFFAOYSA-N
Synonym 1-2-hydroxy-4-methyl-phenyl-ethanone
PubChem CID 81338
IUPAC Name 1-(2-hydroxy-4-methylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09999DA View Documents Fisher Scientific Company
BTB09999DA
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6921-64-8
150.177
1-2-hydroxy-4-methyl-phenyl-ethanone
1-(2-hydroxy-4-methylphenyl)ethanone
C9H10O2
LYKDOWJROLHYOT-UHFFFAOYSA-N
81338
CC1=CC(=C(C=C1)C(=O)C)O

Specifications

1g
96.5
97%
1-2-hydroxy-4-methyl-phenyl-ethanone
CC1=CC(=C(C=C1)C(=O)C)O
150.177
150.18
1-(2-Hydroxy-4-methylphenyl)ethan-1-one
6921-64-8
100.0
C9H10O2
LYKDOWJROLHYOT-UHFFFAOYSA-N
1-(2-hydroxy-4-methylphenyl)ethanone
81338
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning