missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 25790-35-6 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00041759 |
InChI Key | GPYKJDYMMUIUFG-UHFFFAOYSA-N |
Synonym | 1,3-butanedione, 1-2-furanyl, 1-2-furanyl-1,3-butanedione, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-1,3-butanedione, 1-2-furyl-2,3-butanedione, 1-furan-2-yl butane-1,3-dione, 2-furoylacetone, furfuryl-1,3-butanedione, furoyl acetone |
PubChem CID | 12168 |
IUPAC Name | 1-(furan-2-yl)butane-1,3-dione |
SMILES | CC(=O)CC(=O)C1=CC=CO1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC410830250
|
Thermo Fisher Scientific
AC410830250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
25790-35-6 | |
152.15 | |
GPYKJDYMMUIUFG-UHFFFAOYSA-N | |
12168 | |
CC(=O)CC(=O)C1=CC=CO1 |
C8H8O3 | |
MFCD00041759 | |
1,3-butanedione, 1-2-furanyl, 1-2-furanyl-1,3-butanedione, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-1,3-butanedione, 1-2-furyl-2,3-butanedione, 1-furan-2-yl butane-1,3-dione, 2-furoylacetone, furfuryl-1,3-butanedione, furoyl acetone | |
1-(furan-2-yl)butane-1,3-dione |
Specifications
25790-35-6 | |
100.0 | |
98.0°C (5.0 mmHg) | |
95% min. (GC) | |
C8H8O3 | |
Glass bottle | |
1,3-butanedione, 1-2-furanyl, 1-2-furanyl-1,3-butanedione, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-1,3-butanedione, 1-2-furyl-2,3-butanedione, 1-furan-2-yl butane-1,3-dione, 2-furoylacetone, furfuryl-1,3-butanedione, furoyl acetone | |
GPYKJDYMMUIUFG-UHFFFAOYSA-N | |
1-(furan-2-yl)butane-1,3-dione | |
12168 | |
Liquid After Melting | |
1-(2-Furyl)-1, 3-butanedione |
95.0 | |
Undesignated | |
>110°C | |
Authentic | |
MFCD00041759 | |
1.5735 to 1.5755 | |
25g | |
CC(=O)CC(=O)C1=CC=CO1 | |
152.15 | |
152.15 | |
96% |
Safety and Handling
EINECSNumber : 247-265-
RUO – Research Use Only