missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific™

$107.11 - $322.23

Chemical Identifiers

CAS 25790-35-6
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00041759
InChI Key GPYKJDYMMUIUFG-UHFFFAOYSA-N
Synonym 1-2-furyl-1,3-butanedione, 2-furoylacetone, 1-furan-2-yl butane-1,3-dione, 1-2-furanyl-1,3-butanedione, 1,3-butanedione, 1-2-furanyl, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-2,3-butanedione, furoyl acetone, furfuryl-1,3-butanedione
PubChem CID 12168
IUPAC Name 1-(furan-2-yl)butane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2404503
View Documents Promotion Details
Thermo Scientific Chemicals
B2404503
1 g
Each for $107.11
 
AAB2404506
View Documents Promotion Details
Thermo Scientific Chemicals
B2404506
5 g
Each for $322.23
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

25790-35-6
152.149
GPYKJDYMMUIUFG-UHFFFAOYSA-N
12168
CC(=O)CC(=O)C1=CC=CO1
C8H8O3
MFCD00041759
1-2-furyl-1,3-butanedione, 2-furoylacetone, 1-furan-2-yl butane-1,3-dione, 1-2-furanyl-1,3-butanedione, 1,3-butanedione, 1-2-furanyl, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-2,3-butanedione, furoyl acetone, furfuryl-1,3-butanedione
1-(furan-2-yl)butane-1,3-dione

Specifications

25790-35-6
C8H8O3
1 g
GPYKJDYMMUIUFG-UHFFFAOYSA-N
1-(furan-2-yl)butane-1,3-dione
12168
96%
84°C to 86°C (1mmHg)
MFCD00041759
1-2-furyl-1,3-butanedione, 2-furoylacetone, 1-furan-2-yl butane-1,3-dione, 1-2-furanyl-1,3-butanedione, 1,3-butanedione, 1-2-furanyl, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-2,3-butanedione, furoyl acetone, furfuryl-1,3-butanedione
CC(=O)CC(=O)C1=CC=CO1
152.149
152.15
1-(2-Furyl)-1,3-butanedione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 247-265-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'