Please login to your online account to display your discounted pricing

1-(2-Furyl)-1,3-butanedione, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC410830250

 View more versions of this product

Catalog No. AC410830250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid After Melting
Formula Weight 152.15
Quantity 25g
Color Clear yellow to orange-brown
CAS Min % 95.0
CAS Max % 100.0
Molecular Formula C8H8O3
Chemical Name or Material 1-(2-Furyl)-1, 3-butanedione
Assay Percent Range 96%
CAS 25790-35-6
MDL Number MFCD00041759
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-(2-Furyl)-1, 3-butanedione
CAS 25790-35-6
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00041759