Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Furoyl)piperazine, 98%

$103.07 - $1,383.20

Chemical Identifiers

Chemical Name or Material 1-(2-Furoyl)piperazine
CAS 40172-95-0
Molecular Formula C9H12N2O2
Formula Weight 180.21
MDL Number MFCD00038831
Beilstein 880196
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1677314

 
alfa aesar™
A1677314
25g Each for $103.07

AAA1677336

 
alfa aesar™
A1677336
500g Each for $1,383.20

AAA1677322

 
alfa aesar™
A1677322
100g Each for $319.43
Description & Specifications

Specifications

Formula Weight 180.21
Assay 98%
MDL Number MFCD00038831
Boiling Point 137° to 140°C (0.1mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Beilstein 880196
Molecular Formula C9H12N2O2
CAS 40172-95-0
Health Hazard 1 Warning
Chemical Name or Material 1-(2-Furoyl)piperazine
Melting Point 66° to 70°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.