Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluorophenyl)pyrrole-2-carboxaldehyde, 97%

$171.88 - $1,293.00

Chemical Identifiers

Chemical Name or Material 1-(2-Fluorophenyl)pyrrole-2-carboxaldehyde
CAS 132407-65-9
Molecular Formula C11H8FNO
Formula Weight 189.19g/mol
MDL Number MFCD01443629
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5046003

 
alfa aesar™
H50460-03
1g Each for $171.88

AAH5046006

 
alfa aesar™
H50460-06
5g Each for $1,293.00
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 1-(2-Fluorophenyl)pyrrole-2-carboxaldehyde
CAS 132407-65-9
Assay 97%
Molecular Formula C11H8FNO
Formula Weight 189.19g/mol
MDL Number MFCD01443629
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.