missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Fluorophenyl)piperazine Hydrochloride, TCI America™

Manufacturer:  TCI America F029125G

 View more versions of this product

Catalog No. F029125G



Specifications

Specifications

1-(2-Fluorophenyl)piperazine Hydrochloride
190°C
1011-16-1
MFCD00134409
JVQOFQHGEFLRRQ-UHFFFAOYSA-N
1-(2-fluorophenyl)piperazine;hydrochloride
2734698
Crystal-Powder at 20°C
White-Yellow
25g
C10H14ClFN2
1-2-fluorophenyl piperazine hydrochloride, n-2-fluorophenyl piperazine hydrochloride, 1-2-fluorophenyl piperazine monohydrochloride, 1-2-fluorophenyl piperazine hcl, pubchem15282, timtec-bb sbb003382, acmc-2097ud, labotest-bb lt00160127, ksc495m4b, 2-fluorophenyl piperazine, chloride
C1CN(CCN1)C2=CC=CC=C2F.Cl
216.684
216.68
≥98.0% (T)

Chemical Identifiers

1011-16-1
216.684
JVQOFQHGEFLRRQ-UHFFFAOYSA-N
2734698
C1CN(CCN1)C2=CC=CC=C2F.Cl
C10H14ClFN2
MFCD00134409
1-2-fluorophenyl piperazine hydrochloride, n-2-fluorophenyl piperazine hydrochloride, 1-2-fluorophenyl piperazine monohydrochloride, 1-2-fluorophenyl piperazine hcl, pubchem15282, timtec-bb sbb003382, acmc-2097ud, labotest-bb lt00160127, ksc495m4b, 2-fluorophenyl piperazine, chloride
1-(2-fluorophenyl)piperazine;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents