Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluorophenyl)piperazine, 99%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluorophenyl)piperazine
CAS 1011-15-0
Molecular Formula C10H13FN2
Formula Weight 180.23
MDL Number MFCD00040726
UN Number UN2923
Beilstein 611787
Synonym 2-(1-Piperazinyl)fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1939506

 
alfa aesar™
L1939506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.23
Assay 99%
Molecular Formula C10H13FN2
MDL Number MFCD00040726
CAS 1011-15-0
UN Number UN2923
Beilstein 611787
Synonym 2-(1-Piperazinyl)fluorobenzene
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Boiling Point 150°C (3mmHg)
Chemical Name or Material 1-(2-Fluorophenyl)piperazine
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flash Point >110°C (230°F)
Melting Point 44° to 48°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.