missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Fluorophenyl)piperazine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268690100

 View more versions of this product

Catalog No. AC268690100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-(2-Fluorophenyl)piperazine
Authentic
1.5550 to 1.558
Colorless to Yellow
97%
96.0
96% min. (GC)
MFCD00040726
IVTZRJKKXSKXKO-UHFFFAOYSA-N
1-(2-fluorophenyl)piperazine
70529
Liquid
1.1410g/mL
Plastic bottle
1.141
10g
1011-15-0
100.0
C10H13FN2
1-2-fluoro-phenyl-piperazine, 1-2-fluorophenyl piperazine, 1-2-fluorophenyl piperazinehydrochloride, 1-2-fluorophenyl-piperazine, 2-fluorophenyl piperazine, 2-fluorophenylpiperazine, chembl9429, ofpp, piperazine, 1-2-fluorophenyl, pubchem8573
C1CN(CCN1)C2=CC=CC=C2F
180.22
180.22
97%

Chemical Identifiers

1011-15-0
180.22
IVTZRJKKXSKXKO-UHFFFAOYSA-N
70529
C1CN(CCN1)C2=CC=CC=C2F
C10H13FN2
MFCD00040726
1-2-fluoro-phenyl-piperazine, 1-2-fluorophenyl piperazine, 1-2-fluorophenyl piperazinehydrochloride, 1-2-fluorophenyl-piperazine, 2-fluorophenyl piperazine, 2-fluorophenylpiperazine, chembl9429, ofpp, piperazine, 1-2-fluorophenyl, pubchem8573
1-(2-fluorophenyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 213-780-4

TSCA : TSCA

SDS Documents