missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Fluorophenyl)piperazine, 97%, ACROS Organics™

$296.27 - $296.27

Chemical Identifiers

CAS 1011-15-0
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.22
MDL Number MFCD00040726
InChI Key IVTZRJKKXSKXKO-UHFFFAOYSA-N
Synonym 1-2-fluoro-phenyl-piperazine, 1-2-fluorophenyl piperazine, 1-2-fluorophenyl piperazinehydrochloride, 1-2-fluorophenyl-piperazine, 2-fluorophenyl piperazine, 2-fluorophenylpiperazine, chembl9429, ofpp, piperazine, 1-2-fluorophenyl, pubchem8573
PubChem CID 70529
IUPAC Name 1-(2-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=CC=C2F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC268690100
SDS View Documents
Thermo Fisher Scientific
268690100
10g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC268691000
SDS View Documents
Thermo Fisher Scientific
268691000
100g Plastic bottle
Each for $296.27
 
Description

Description

Specifications

Chemical Identifiers

1011-15-0
180.22
IVTZRJKKXSKXKO-UHFFFAOYSA-N
70529
C1CN(CCN1)C2=CC=CC=C2F
C10H13FN2
MFCD00040726
1-2-fluoro-phenyl-piperazine, 1-2-fluorophenyl piperazine, 1-2-fluorophenyl piperazinehydrochloride, 1-2-fluorophenyl-piperazine, 2-fluorophenyl piperazine, 2-fluorophenylpiperazine, chembl9429, ofpp, piperazine, 1-2-fluorophenyl, pubchem8573
1-(2-fluorophenyl)piperazine

Specifications

1.1410g/mL
Plastic bottle
1.141
10g
96.0
96% min. (GC)
MFCD00040726
IVTZRJKKXSKXKO-UHFFFAOYSA-N
1-(2-fluorophenyl)piperazine
70529
Liquid
1-(2-Fluorophenyl)piperazine, 97%
Authentic
1.5550 to 1.558
Colorless to Yellow
1011-15-0
100.0
C10H13FN2
1-2-fluoro-phenyl-piperazine, 1-2-fluorophenyl piperazine, 1-2-fluorophenyl piperazinehydrochloride, 1-2-fluorophenyl-piperazine, 2-fluorophenyl piperazine, 2-fluorophenylpiperazine, chembl9429, ofpp, piperazine, 1-2-fluorophenyl, pubchem8573
C1CN(CCN1)C2=CC=CC=C2F
180.22
180.22
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 213-780-4

TSCA : TSCA

SDS