missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluorobenzoyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H5633903

 View more versions of this product

Catalog No. AAH5633903



Description

Description

Applications
2‘-fluoro-6-pyrrol-2-phenyl-4-quinolone, a derivative of 1-(2-Fluorobenzoyl)pyrrolidine exhibited the most potent cytotoxic activities against renal and melanoma tumor cell lines.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

1-(2-Fluorobenzoyl)pyrrolidine
347909-09-5
MFCD00460952
AZBXHPRYPHBYCV-UHFFFAOYSA-N
(2-fluorophenyl)-pyrrolidin-1-ylmethanone
3497148
97%
1g
C11H12FNO
1-2-fluorobenzoyl pyrrolidine, 2-fluorophenyl 1-pyrrolidinyl methanone, 2-fluorophenyl-pyrrolidin-1-ylmethanone
C1CCN(C1)C(=O)C2=CC=CC=C2F
193.221
193.22

Chemical Identifiers

347909-09-5
193.221
AZBXHPRYPHBYCV-UHFFFAOYSA-N
3497148
C1CCN(C1)C(=O)C2=CC=CC=C2F
C11H12FNO
MFCD00460952
1-2-fluorobenzoyl pyrrolidine, 2-fluorophenyl 1-pyrrolidinyl methanone, 2-fluorophenyl-pyrrolidin-1-ylmethanone
(2-fluorophenyl)-pyrrolidin-1-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents