Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)pyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)pyrrolidine
CAS 1233952-03-8
Formula Weight 210.2
MDL Number MFCD13689280
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6307103

 
alfa aesar™
H6307103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6307106

 
alfa aesar™
H6307106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63071MD

 
alfa aesar™
H63071MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.2
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)pyrrolidine
Physical Form Solid
Assay 97%
MDL Number MFCD13689280
CAS 1233952-03-8
Quantity 1g