missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)piperidine, 97%

Manufacturer:  Alfa Aesar™ H6346903

 View more versions of this product

Catalog No. AAH6346903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-(2-Fluoro-6-nitrophenyl)piperidine
1233952-03-8
MFCD13689258
QJQZIIGCNJGTMH-UHFFFAOYSA-N
1-(2-fluoro-6-nitrophenyl)pyrrolidine
53255583
Solid
1g
C10H11FN2O2
1-2-fluoro-6-nitrophenyl pyrrolidine
C1CCN(C1)C2=C(C=CC=C2F)[N+](=O)[O-]
210.208
224.23
97%

Chemical Identifiers

1233952-03-8
210.208
QJQZIIGCNJGTMH-UHFFFAOYSA-N
53255583
C1CCN(C1)C2=C(C=CC=C2F)[N+](=O)[O-]
C10H11FN2O2
MFCD13689258
1-2-fluoro-6-nitrophenyl pyrrolidine
1-(2-fluoro-6-nitrophenyl)pyrrolidine
Documents