Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)piperidine
Formula Weight 224.23
MDL Number MFCD13689258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6346903

 
alfa aesar™
H6346903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6346906

 
alfa aesar™
H6346906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63469MD

 
alfa aesar™
H63469MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.23
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)piperidine
Physical Form Solid
Assay 97%
MDL Number MFCD13689258
Quantity 1g