Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)piperazine
CAS 1234010-04-8
Formula Weight 225.22
Beilstein 19933446
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6307703

 
alfa aesar™
H6307703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6307706

 
alfa aesar™
H6307706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63077MD

 
alfa aesar™
H63077MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.22
CAS 1234010-04-8
Assay 97%
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)piperazine
Physical Form Solid
Quantity 1g
Beilstein 19933446