Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)-4-piperidone, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-piperidone
Formula Weight 238.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6322603

 
alfa aesar™
H6322603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6322606

 
alfa aesar™
H6322606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63226MD

 
alfa aesar™
H63226MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.22
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-piperidone
Physical Form Solid
Assay 97%
Quantity 1g