Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine
Formula Weight 239.25
MDL Number MFCD13689279
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6317503

 
alfa aesar™
H6317503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6317506

 
alfa aesar™
H6317506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63175MD

 
alfa aesar™
H63175MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.25
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine
Physical Form Solid
Assay 97%
MDL Number MFCD13689279
Quantity 1g