missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)pyrrolidine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 385380-74-5
Molecular Formula C10H11FN2O2
Molecular Weight (g/mol) 210.208
InChI Key SMKXLBISYVJESG-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl pyrrolidine, 3-fluoro-4-pyrrolidin-1-yl nitrobenzene, pyrrolidine, 1-2-fluoro-4-nitrophenyl, pubchem18700, cambridge id 5842403, 2-fluoro-4-nitrophenyl pyrrolidine, 1-2-fluoro-4-nitro-phenyl pyrrolidine
PubChem CID 2872122
IUPAC Name 1-(2-fluoro-4-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63919MD View Documents Alfa Aesar™
H63919MD
250mg Each for $35.93
 
AAH6391903 View Documents Alfa Aesar™
H6391903
1g Each for $106.26
 
AAH6391906 View Documents Alfa Aesar™
H6391906
5g Each for $422.54
 
Specifications

Chemical Identifiers

385380-74-5
210.208
1-2-fluoro-4-nitrophenyl pyrrolidine, 3-fluoro-4-pyrrolidin-1-yl nitrobenzene, pyrrolidine, 1-2-fluoro-4-nitrophenyl, pubchem18700, cambridge id 5842403, 2-fluoro-4-nitrophenyl pyrrolidine, 1-2-fluoro-4-nitro-phenyl pyrrolidine
1-(2-fluoro-4-nitrophenyl)pyrrolidine
C10H11FN2O2
SMKXLBISYVJESG-UHFFFAOYSA-N
2872122
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

250mg
C10H11FN2O2
SMKXLBISYVJESG-UHFFFAOYSA-N
1-(2-fluoro-4-nitrophenyl)pyrrolidine
2872122
Solid
1-(2-Fluoro-4-nitrophenyl)pyrrolidine
385380-74-5
1-2-fluoro-4-nitrophenyl pyrrolidine, 3-fluoro-4-pyrrolidin-1-yl nitrobenzene, pyrrolidine, 1-2-fluoro-4-nitrophenyl, pubchem18700, cambridge id 5842403, 2-fluoro-4-nitrophenyl pyrrolidine, 1-2-fluoro-4-nitro-phenyl pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F
210.208
210.2
97%