Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)piperidine
CAS 172967-04-3
Formula Weight 224.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6326703

 
alfa aesar™
H6326703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6326706

 
alfa aesar™
H6326706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63267MD

 
alfa aesar™
H63267MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.23
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)piperidine
Assay 97%
CAS 172967-04-3
Quantity 1g
Physical Form Solid