Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)piperazine
CAS 154590-33-7
Formula Weight 225.22
MDL Number MFCD01572463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6305903

 
alfa aesar™
H6305903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6305906

 
alfa aesar™
H6305906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63059MD

 
alfa aesar™
H63059MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.22
Assay 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)piperazine
Quantity 1g
MDL Number MFCD01572463
CAS 154590-33-7