Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)azepane, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)azepane
CAS 250371-80-3
Formula Weight 238.25
UN Number UN3259
Synonym 1-(2-Fluoro-4-nitrophenyl)hexamethyleneimine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6330503

  SDS
alfa aesar™
H6330503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6330506

  SDS
alfa aesar™
H6330506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63305MD

  SDS
alfa aesar™
H63305MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.25
UN Number UN3259
Synonym 1-(2-Fluoro-4-nitrophenyl)hexamethyleneimine
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Quantity 1g
CAS 250371-80-3
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)azepane
Assay 97%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.