missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)-4-piperidone, 97%

$71.08 - $845.08

Chemical Identifiers

CAS 439097-58-2
Molecular Formula C11H11FN2O3
Molecular Weight (g/mol) 238.218
InChI Key BMXFDVAJYBTGRC-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl piperidin-4-one, 1-2-fluoro-4-nitrophenyl tetrahydro-4 1h-pyridinone, 1-2-fluoro-4-nitro-phenyl-piperidin-4-one, 4-piperidinone, 1-2-fluoro-4-nitrophenyl, 4-4-oxo-piperidin-1-yl-3-fluoronitrobenzene
PubChem CID 3763781
IUPAC Name 1-(2-fluoro-4-nitrophenyl)piperidin-4-one
SMILES C1CN(CCC1=O)C2=C(C=C(C=C2)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63489MD View Documents Alfa Aesar™
H63489MD
250mg Each for $71.08
 
AAH6348903 View Documents Alfa Aesar™
H6348903
1g Each for $211.27
 
AAH6348906 View Documents Alfa Aesar™
H6348906
5g Each for $845.08
 
Specifications

Chemical Identifiers

439097-58-2
238.218
1-2-fluoro-4-nitrophenyl piperidin-4-one, 1-2-fluoro-4-nitrophenyl tetrahydro-4 1h-pyridinone, 1-2-fluoro-4-nitro-phenyl-piperidin-4-one, 4-piperidinone, 1-2-fluoro-4-nitrophenyl, 4-4-oxo-piperidin-1-yl-3-fluoronitrobenzene
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
C11H11FN2O3
BMXFDVAJYBTGRC-UHFFFAOYSA-N
3763781
C1CN(CCC1=O)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

250mg
C11H11FN2O3
1-2-fluoro-4-nitrophenyl piperidin-4-one, 1-2-fluoro-4-nitrophenyl tetrahydro-4 1h-pyridinone, 1-2-fluoro-4-nitro-phenyl-piperidin-4-one, 4-piperidinone, 1-2-fluoro-4-nitrophenyl, 4-4-oxo-piperidin-1-yl-3-fluoronitrobenzene
C1CN(CCC1=O)C2=C(C=C(C=C2)[N+](=O)[O-])F
238.218
238.22
97%
439097-58-2
14265568
BMXFDVAJYBTGRC-UHFFFAOYSA-N
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
3763781
Solid
1-(2-Fluoro-4-nitrophenyl)-4-piperidone