Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine
CAS 221198-29-4
Formula Weight 239.25
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6376203

 
alfa aesar™
H6376203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6376206

 
alfa aesar™
H6376206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63762MD

 
alfa aesar™
H63762MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.25
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine
Physical Form Solid
Assay 97%
CAS 221198-29-4
Quantity 1g