Please login to your online account to display your discounted pricing

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01820DA

 View more versions of this product

Catalog No. SB01820DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 228.26
Quantity 1g
Assay 97%
CAS Min % 97.0
MDL Number MFCD00236612
CAS 37989-92-7
Chemical Name or Material 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
Assay Percent Range 97%
Molecular Formula C15H13FO
Linear Formula FC6H4C6H4C(O)CH2CH3
Packaging Amber glass bottle

Specifications

Chemical Name or Material 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
CAS 37989-92-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H13FO
Linear Formula FC6H4C6H4C(O)CH2CH3
Formula Weight 228.26
MDL Number MFCD00236612