Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Ethanedisulfonic acid hydrate, 98%

Chemical Identifiers

Chemical Name or Material 1,2-Ethanedisulfonic acid hydrate
CAS 110-04-3
Molecular Formula HO3SCH2CH2SO3H·xH2O
Formula Weight 190.20 (Anhydrous)
MDL Number MFCD00069901
UN Number UN3261
Merck Index 14,3724
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2095206

  SDS
alfa aesar™
B2095206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2095214

  SDS
alfa aesar™
B2095214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2095222

  SDS
alfa aesar™
B2095222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.20 (Anhydrous)
Molecular Formula HO3SCH2CH2SO3H·xH2O
MDL Number MFCD00069901
Merck Index 14,3724
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3261
Chemical Name or Material 1,2-Ethanedisulfonic acid hydrate
CAS 110-04-3
Assay 98%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Quantity 5g
Health Hazard 1 Danger

Applications
It is employed in the preparation of “edisylate“ salts. It is also used in biological studies to evaluate cellular metabolism and allergy using guinea pigs as the subject of the studies.

Solubility
Soluble in water, ethanol, ether, and very soluble in dioxane.

Notes
Store away from oxidizing agents, strong bases and water or moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.