missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Ethanedisulfonic Acid Disodium Salt, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 409461000

 View more versions of this product

Catalog No. AC409461000


 

Specifications Description & Specifications

Specifications

≥96%
Crystalline Powder
White
Authentic
sodium bromide content (by titration): < 1.5%
5325-43-9
NaO3SCH2CH2SO3Na
234.16
100g
3% max. (135°C)
Plastic bottle
1, 2-Ethanedisulfonic acid, disodium salt
MFCD00024998
C2H4Na2O6S2

Chemical Identifiers

C2H4Na2O6S2
234.16
1, 2-Ethanedisulfonic acid, disodium salt
NaO3SCH2CH2SO3Na
MFCD00024998
≥96%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-198-0

TSCA : TSCA