missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Ethanedisulfonic Acid Disodium Salt, ACROS Organics™

$437.27 - $437.27

Chemical Identifiers

Molecular Formula C2H4Na2O6S2
Linear Formula NaO3SCH2CH2SO3Na
Formula Weight 234.16
MDL Number MFCD00024998
Chemical Name or Material 1, 2-Ethanedisulfonic acid, disodium salt
Assay ≥96%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC409460050 SDS Thermo Fisher Scientific
409460050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC409461000 SDS Thermo Fisher Scientific
409461000
Plastic bottle 100g Each for $437.27
Specifications Description & Specifications

Chemical Identifiers

C2H4Na2O6S2
234.16
1, 2-Ethanedisulfonic acid, disodium salt
NaO3SCH2CH2SO3Na
MFCD00024998
≥96%

Specifications

sodium bromide content (by titration): < 1.5%
Crystalline Powder
NaO3SCH2CH2SO3Na
234.16
3% max. (135°C)
White
1, 2-Ethanedisulfonic acid, disodium salt
5325-43-9
C2H4Na2O6S2
Authentic
MFCD00024998
Glass bottle
5g
≥96%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-198-0

TSCA : TSCA