Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Ethanedisulfonic acid disodium salt, 99%

$107.08 - $107.08

Chemical Identifiers

Chemical Name or Material 1,2-Ethanedisulfonic acid disodium salt
CAS 5325-43-9
Molecular Formula NaO3SCH2CH2SO3Na
Formula Weight 234.16g/mol
MDL Number MFCD00024998
Beilstein 3920126
Merck Index 14,3724
Synonym Disodium 1,2-ethanedisulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2079314

 
alfa aesar™
B20793-14
25g Each for $107.08
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material 1,2-Ethanedisulfonic acid disodium salt
CAS 5325-43-9
Assay 99%
Molecular Formula NaO3SCH2CH2SO3Na
Formula Weight 234.16g/mol
MDL Number MFCD00024998
Beilstein 3920126
Merck Index 14,3724
Synonym Disodium 1,2-ethanedisulfonate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA