missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Ethanedisulfonic acid dihydrate 98%, ACROS Organics™

Chemical Identifiers

CAS 5982-56-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C2H6O6S2·2H2O
Formula Weight 226.22
MDL Number MFCD00069901
Chemical Name or Material 1, 2-Ethanedisulfonic acid dihydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC384800050 SDS Thermo Fisher Scientific
AC384800050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC384800250 SDS Thermo Fisher Scientific
AC384800250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5982-56-9
100.0
C2H6O6S2·2H2O
MFCD00069901
97.5
98%
226.22
1, 2-Ethanedisulfonic acid dihydrate

Specifications

226.22
15, 3779
100.0
MFCD00069901
97.5
C2H6O6S2·2H2O
5g
04, IV, 78
5982-56-9
98%
Glass bottle
1, 2-Ethanedisulfonic acid dihydrate

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger