missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Ethanedisulfonic acid dihydrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 384800250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 2-Ethanedisulfonic acid dihydrate | |
97.5 | |
98% | |
C2H6O6S2·2H2O | |
25g | |
15, 3779 | |
Solubility in water: very soluble | |
C(CS(=O)(=O)O)S(=O)(=O)O.O.O | |
226.22 | |
226.22 |
5982-56-9 | |
100.0 | |
Glass bottle | |
MFCD00069901 | |
04,II,528 | |
1,2-ethanedisulfonic acid dihydrate, unii-0j653yp17s, 1,2-ethanedisulfonic acid, dihydrate, 1,2-ethanedisulfonic acid dihydrate mi, 1,2-ethanedisulfonic acid, hydrate 1:2, ethane disulfonic acid dihydrate, ethane-1,2-disulfonic acid dihydrate, ethane-1,2-disulfonic acid-water 1/2 | |
DSGUSEBCDAKBCM-UHFFFAOYSA-N | |
ethane-1,2-disulfonic acid;dihydrate | |
57346009 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO â Research Use Only