missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxypentane, TCI America™

Manufacturer:  TCI America E082225ML

 View more versions of this product

Catalog No. E082225ML



Specifications

Specifications

1,2-Epoxypentane
Colorless
1003-14-1
MFCD00192124
1,2-Pentylene Oxide, Propyloxirane
CCCC1CO1
86.134
86.13
≥98.0% (GC)
90°C
25mL
C5H10O
1993
SYURNNNQIFDVCA-UHFFFAOYSA-N
2-propyloxirane
13853
Clear Liquid at 20°C

Chemical Identifiers

1003-14-1
86.134
SYURNNNQIFDVCA-UHFFFAOYSA-N
13853
CCCC1CO1
C5H10O
MFCD00192124
1,2-Pentylene Oxide, Propyloxirane
2-propyloxirane
Safety and Handling

Safety and Handling

RTECSNumber : SA2700000

TSCA : Yes

Documents