Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Epoxyoctane, 97%

Chemical Identifiers

Chemical Name or Material 1,2-Epoxyoctane
CAS 2984-50-1
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00005157
UN Number UN1993
Beilstein 79912
Synonym 1-Octene oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2146314

  SDS
alfa aesar™
B2146314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2146322

  SDS
alfa aesar™
B2146322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.22
Chemical Name or Material 1,2-Epoxyoctane
Assay 97%
Molecular Formula C8H16O
MDL Number MFCD00005157
CAS 2984-50-1
UN Number UN1993
Beilstein 79912
Synonym 1-Octene oxide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 167°C
Quantity 25g
Flash Point 37°C (98°F)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number RG9625000

Applications
It is employed as an intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C