missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxyoctadecane, TCI America™

Manufacturer:  TCI America E031325G

 View more versions of this product

Catalog No. E031325G



Specifications

Specifications

1,2-Epoxyoctadecane
White-Yellow
7390-81-0
MFCD00005152
QBJWYMFTMJFGOL-UHFFFAOYSA-N
2-hexadecyloxirane
23872
Crystal-Lump at 20°C
137°C
25g
C18H36O
1,2-epoxyoctadecane, hexadecyloxirane, oxirane, hexadecyl, octadecane, epoxy, 1,2-octadecylene oxide, ccris 2619, oxirane, 2-hexadecyl, #, 1,2-epoxy octadecane, acmc-209osu
CCCCCCCCCCCCCCCCC1CO1
268.485
268.49
≥85.0% (GC)

Chemical Identifiers

7390-81-0
268.485
QBJWYMFTMJFGOL-UHFFFAOYSA-N
23872
CCCCCCCCCCCCCCCCC1CO1
C18H36O
MFCD00005152
1,2-epoxyoctadecane, hexadecyloxirane, oxirane, hexadecyl, octadecane, epoxy, 1,2-octadecylene oxide, ccris 2619, oxirane, 2-hexadecyl, #, 1,2-epoxy octadecane, acmc-209osu
2-hexadecyloxirane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0220

TSCA : Yes

Documents