Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Epoxydodecane, 95%

$55.76 - $174.78

Chemical Identifiers

Chemical Name or Material 1,2-Epoxydodecane
CAS 2855-19-8
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00005150
Beilstein 105569
Synonym 1-Dodecene oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0729514

  SDS
alfa aesar™
L0729514
25g Each for $55.76

AAL0729522

  SDS
alfa aesar™
L0729522
100g Each for $174.78
Description & Specifications

Specifications

Formula Weight 184.32
Molecular Formula C12H24O
Synonym 1-Dodecene oxide
CAS 2855-19-8
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 1,2-Epoxydodecane
Health Hazard 1 Warning
MDL Number MFCD00005150
RTECS Number JR2450000
Boiling Point 121° to 122°C (12mmHg)
Assay 95%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Beilstein 105569
Flash Point 105°C (221°F)
Quantity 25g

Applications
1,2-Epoxydodecane is used as a host material in the synthesis of polymer intercalation. It is used as organic intermediates and in pharmaceutical research and development.

Solubility
Not miscible in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.