missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Epoxydecane, 97%

$41.42 - $153.47

Chemical Identifiers

CAS 2404-44-6
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00005158
Beilstein 104132
Synonym 1-Decene oxide
Chemical Name or Material 1,2-Epoxydecane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0728906 SDS Alfa Aesar™
L0728906
5g Each for $41.42
AAL0728914 SDS Alfa Aesar™
L0728914
25g Each for $153.47
Specifications Description & Specifications

Chemical Identifiers

2404-44-6
156.27
104132
1,2-Epoxydecane
C10H20O
MFCD00005158
1-Decene oxide

Specifications

2404-44-6
156.27
104132
92° to 93°C (14mmHg)
78°C (172°F)
97%
C10H20O
MFCD00005158
1-Decene oxide
5g
1,2-Epoxydecane

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA