missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxycyclohexane, TCI America™

Manufacturer:  TCI America O0071500ML

 View more versions of this product

Catalog No. O0071500ML



Specifications

Specifications

1,2-Epoxycyclohexane
Colorless
286-20-4
MFCD00005162
cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide
C1CCC2C(C1)O2
98.145
98.15
≥98.0% (GC)
130°C
500mL
C6H10O
1993
ZWAJLVLEBYIOTI-UHFFFAOYSA-N
7-oxabicyclo[4.1.0]heptane
9246
Clear Liquid at 20°C

Chemical Identifiers

286-20-4
98.145
ZWAJLVLEBYIOTI-UHFFFAOYSA-N
9246
C1CCC2C(C1)O2
C6H10O
MFCD00005162
cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide
7-oxabicyclo[4.1.0]heptane
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3263

RTECSNumber : RN7175000

TSCA : Yes

Documents