missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxycyclododecane (mixture of isomers), TCI America™

Manufacturer:  TCI America E032225ML

 View more versions of this product

Catalog No. E032225ML



Specifications

Specifications

1,2-Epoxycyclododecane (mixture of isomers)
Yellow
286-99-7
MFCD00005116
VLJLXEKIAALSJE-UHFFFAOYSA-N
13-oxabicyclo[10.1.0]tridecane
9248
Clear Liquid at 20°C
130°C
25mL
C12H22O
1,2-epoxycyclododecane, 13-oxabicyclo 10.1.0 tridecane, epoxycyclododecane, cyclododecene oxide, cyclododecene epoxide, cyclododecane epoxide, cyclododecane, 1,2-epoxy, ccris 3754, 13-oxabicyclo 10.1.0 tridecane, cyclododecane oxide
C1CCCCCC2C(O2)CCCC1
182.307
182.31
≥95.0% (GC)

Chemical Identifiers

286-99-7
182.307
VLJLXEKIAALSJE-UHFFFAOYSA-N
9248
C1CCCCCC2C(O2)CCCC1
C12H22O
MFCD00005116
1,2-epoxycyclododecane, 13-oxabicyclo 10.1.0 tridecane, epoxycyclododecane, cyclododecene oxide, cyclododecene epoxide, cyclododecane epoxide, cyclododecane, 1,2-epoxy, ccris 3754, 13-oxabicyclo 10.1.0 tridecane, cyclododecane oxide
13-oxabicyclo[10.1.0]tridecane
Safety and Handling

Safety and Handling

TSCA : Yes

Documents