missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxy-9-decene, TCI America™

Manufacturer:  TCI America E041125ML

 View more versions of this product

Catalog No. E041125ML



Specifications

Specifications

1,2-Epoxy-9-decene
Colorless
85721-25-1
MFCD00069427
FCZHJHKCOZGQJZ-UHFFFAOYSA-N
2-oct-7-enyloxirane
543758
Clear Liquid at 20°C
80°C
25mL
C10H18O
1,2-epoxy-9-decene, oxirane, 7-octenyl, oct-7-en-1-yloxirane, 2-7-octen-1-yl oxirane, 2-oct-7-en-1-yl oxirane, oct-7-enyloxirane, 7-octenyl oxirane, zlchem 822, acmc-20apkm, 2-7-octenyl oxirane
C=CCCCCCCC1CO1
154.253
154.25
≥96.0% (GC)

Chemical Identifiers

85721-25-1
154.253
FCZHJHKCOZGQJZ-UHFFFAOYSA-N
543758
C=CCCCCCCC1CO1
C10H18O
MFCD00069427
1,2-epoxy-9-decene, oxirane, 7-octenyl, oct-7-en-1-yloxirane, 2-7-octen-1-yl oxirane, 2-oct-7-en-1-yl oxirane, oct-7-enyloxirane, 7-octenyl oxirane, zlchem 822, acmc-20apkm, 2-7-octenyl oxirane
2-oct-7-enyloxirane
Safety and Handling

Safety and Handling

TSCA : No

Documents