missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxy-7-octene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 33514-0050

 View more versions of this product

Catalog No. AC335140050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1, 2-Epoxy-7-octene
19600-63-6
100.0
≥96.0%
C8H14O
126.2
1.4345 to 1.4365 (20°C, 589nm)
0.85g/mL
5g
97%
96.0
Liquid
97%
Authentic
MFCD00005156
Glass bottle
Clear colorless

Chemical Identifiers

19600-63-6
100.0
C8H14O
MFCD00005156
≥96.0%
96.0
97%
126.2
1, 2-Epoxy-7-octene, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-178-7

RTECSNumber : RR0502000

TSCA : TSCA