missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxy-7-octene, 97%, ACROS Organics™

Chemical Identifiers

CAS 19600-63-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H14O
Formula Weight 126.2
MDL Number MFCD00005156
Chemical Name or Material 1, 2-Epoxy-7-octene, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC335140050 SDS Thermo Fisher Scientific
33514-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC335140250 SDS Thermo Fisher Scientific
33514-0250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

19600-63-6
100.0
C8H14O
MFCD00005156
≥96.0%
96.0
97%
126.2
1, 2-Epoxy-7-octene, 97%

Specifications

126.2
5g
0.85g/mL
100.0
Authentic
C8H14O
19600-63-6
1, 2-Epoxy-7-octene, 97%
Liquid
Clear colorless
96.0
Glass bottle
1.4345 to 1.4365 (20°C, 589nm)
MFCD00005156
97%
≥96.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-178-7

RTECSNumber : RR0502000

TSCA : TSCA