missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 98.0+%, TCI America™

Supplier:  TCI America E095625ML

 View more versions of this product

Catalog No. E095625ML



Description

Description

Specifications

Specifications

1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)
-100°C
159°C
MFCD00022356
3271
SLJFKNONPLNAPF-UHFFFAOYSA-N
4-ethenyl-7-oxabicyclo[4.1.0]heptane
7832
≥98.0% (GC)
106-86-5
Colorless
C8H12O
25 mL
4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane
C=CC1CCC2C(C1)O2
124.183
124.18
Liquid

Chemical Identifiers

106-86-5
124.183
SLJFKNONPLNAPF-UHFFFAOYSA-N
7832
C=CC1CCC2C(C1)O2
C8H12O
MFCD00022356
4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane
4-ethenyl-7-oxabicyclo[4.1.0]heptane
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2296

RTECSNumber : RN8770000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions