Please login to your online account to display your discounted pricing

1,2-Epoxy-3-(4-nitrophenoxy)propane, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2-Epoxy-3-(4-nitrophenoxy)propane
Name Note 90%
CAS 5255-75-4
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥85%
Assay Percent Range 90%
Molecular Formula C9 H9 N O4
Formula Weight 195.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC409440050

  SDS
thermo fisher scientific
409440050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥85%
Purity Grade Technical Grade
Physical Form Solid
Quantity 5g
Color Yellow
Beilstein 17, V,3, 15
RTECS Number TZ3650000
EINECS Number 226-057-3
Chemical Name or Material 1, 2-Epoxy-3-(4-nitrophenoxy)propane
TSCA TSCA
CAS Max % 100.0
CAS Min % 88.0
Formula Weight 195.17
MDL Number MFCD00041845
Melting Point 62° to 68°C
Packaging Glass bottle
Name Note 90%
Assay Percent Range 90%
Infrared Spectrum Authentic
CAS 5255-75-4
Molecular Formula C9 H9 N O4
Synonym Glycidyl-4-nitrophenyl ether